Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine, 98%

$105.16 - $364.64

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine
CAS 306977-91-3
Molecular Formula C13H9ClN2O
Formula Weight 244.68
MDL Number MFCD00215274
Synonym 2-Chloro-6-(4-methoxyphenyl)nicotinonitrile; 2-Chloro-6-(4-methoxyphenyl)pyridine-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5052003

  SDS
alfa aesar™
H5052003
1g Each for $364.64

AAH50520MD

  SDS
alfa aesar™
H50520MD
250mg Each for $105.16
Description & Specifications

Specifications

Formula Weight 244.68
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 98%
Synonym 2-Chloro-6-(4-methoxyphenyl)nicotinonitrile; 2-Chloro-6-(4-methoxyphenyl)pyridine-3-carbonitrile
Chemical Name or Material 2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine
Molecular Formula C13H9ClN2O
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD00215274
Melting Point 148°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 306977-91-3
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.