Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyano-6-(4-fluorophenyl)pyridine, 96%

$557.22 - $557.22

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-cyano-6-(4-fluorophenyl)pyridine
CAS 31776-83-7
Molecular Formula C12H6ClFN2
Formula Weight 232.65
MDL Number MFCD00215072
Synonym 2-Chloro-6-(4-fluorophenyl)nicotinonitrile; 2-Chloro-6-(4-fluorophenyl)pyridine-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5051906

  SDS
alfa aesar™
H5051906
5g Each for $557.22
Description & Specifications

Specifications

Formula Weight 232.65
Chemical Name or Material 2-Chloro-3-cyano-6-(4-fluorophenyl)pyridine
Assay 96%
Molecular Formula C12H6ClFN2
MDL Number MFCD00215072
CAS 31776-83-7
Synonym 2-Chloro-6-(4-fluorophenyl)nicotinonitrile; 2-Chloro-6-(4-fluorophenyl)pyridine-3-carbonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.