Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3-cyano-5-phenylpyridine, 98%

$87.28 - $304.78

Chemical Identifiers

Chemical Name or Material 2-Chloro-3-cyano-5-phenylpyridine
CAS 10177-10-3
Molecular Formula C12H7ClN2
Formula Weight 214.66
MDL Number MFCD00231610
Synonym 2-Chloro-5-phenylnicotinonitrile; 2-Chloro-5-phenylpyridine-3-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5052103

 
alfa aesar™
H5052103
1g Each for $304.78

AAH50521MD

 
alfa aesar™
H50521MD
250mg Each for $87.28
Description & Specifications

Specifications

Formula Weight 214.66
Quantity 1g
Chemical Name or Material 2-Chloro-3-cyano-5-phenylpyridine
Assay 98%
Molecular Formula C12H7ClN2
MDL Number MFCD00231610
CAS 10177-10-3
Synonym 2-Chloro-5-phenylnicotinonitrile; 2-Chloro-5-phenylpyridine-3-carbonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 143° to 147°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.