missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,6-difluorobenzoyl chloride, 97%

$74.80 - $252.91

Chemical Identifiers

CAS 261762-42-9
Molecular Formula C7H2Cl2F2O
Molecular Weight (g/mol) 210.989
MDL Number MFCD01631442
InChI Key CBFPZKKUVNEIFF-UHFFFAOYSA-N
Synonym 2-chloro-3,6-difluorobenzoylchloride, acmc-20ao08, 2-chloro-3,6,-difluorobenzoyl chloride, benzoyl chloride,2-chloro-3,6-difluoro, 2-chloranyl-3,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 2-chloro-3,6-difluoro-9ci
PubChem CID 2773536
IUPAC Name 2-chloro-3,6-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1F)C(=O)Cl)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1816903 View Documents Alfa Aesar™
L1816903
1g Each for $74.80
 
AAL1816906 View Documents Alfa Aesar™
L1816906
5g Each for $252.91
 
Specifications

Chemical Identifiers

261762-42-9
210.989
CBFPZKKUVNEIFF-UHFFFAOYSA-N
2773536
C1=CC(=C(C(=C1F)C(=O)Cl)Cl)F
C7H2Cl2F2O
MFCD01631442
2-chloro-3,6-difluorobenzoylchloride, acmc-20ao08, 2-chloro-3,6,-difluorobenzoyl chloride, benzoyl chloride,2-chloro-3,6-difluoro, 2-chloranyl-3,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 2-chloro-3,6-difluoro-9ci
2-chloro-3,6-difluorobenzoyl chloride

Specifications

1g
C7H2Cl2F2O
UN3265
CBFPZKKUVNEIFF-UHFFFAOYSA-N
2-chloro-3,6-difluorobenzoyl chloride
2773536
97%
261762-42-9
MFCD01631442
2-chloro-3,6-difluorobenzoylchloride, acmc-20ao08, 2-chloro-3,6,-difluorobenzoyl chloride, benzoyl chloride,2-chloro-3,6-difluoro, 2-chloranyl-3,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 2-chloro-3,6-difluoro-9ci
C1=CC(=C(C(=C1F)C(=O)Cl)Cl)F
210.989
211
2-Chloro-3,6-difluorobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.