Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,6-difluorobenzamide, 97+%

$90.50 - $321.72

Chemical Identifiers

Chemical Name or Material 2-Chloro-3,6-difluorobenzamide
CAS 261762-40-7
Molecular Formula C7H4ClF2NO
Formula Weight 191.56
MDL Number MFCD01631444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2451003

 
alfa aesar™
B2451003
1g Each for $90.50

AAB2451006

 
alfa aesar™
B2451006
5g Each for $321.72
Description & Specifications

Specifications

Formula Weight 191.56
Chemical Name or Material 2-Chloro-3,6-difluorobenzamide
Assay ≥97%
Molecular Formula C7H4ClF2NO
MDL Number MFCD01631444
CAS 261762-40-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.