Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,5-dinitropyridine, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3,5-dinitropyridine
CAS 2578-45-2
Molecular Formula C5H2ClN3O4
Formula Weight 203.54
MDL Number MFCD00006233
UN Number UN2811
Beilstein 196026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0077003

  SDS
alfa aesar™
L0077003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0077006

  SDS
alfa aesar™
L0077006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0077014

  SDS
alfa aesar™
L0077014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.54
Chemical Name or Material 2-Chloro-3,5-dinitropyridine
MDL Number MFCD00006233
CAS 2578-45-2
TSCA TSCA
Molecular Formula C5H2ClN3O4
UN Number UN2811
Beilstein 196026
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number US6504750
Melting Point 60° to 66°C
Quantity 1g
Assay ≥98%
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
2-Chloro-3,5-dinitropyridine is used as efficient pie electron acceptor in the preparation of charge transfer complexes with aniline and its derivatives acting as donors.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.