Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,5-dinitrobenzotrifluoride, 98+%

Chemical Identifiers

Chemical Name or Material 2-Chloro-3,5-dinitrobenzotrifluoride
CAS 392-95-0
Molecular Formula C7H2ClF3N2O4
Formula Weight 270.55
MDL Number MFCD00007076
UN Number UN2923
Beilstein 2061156
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1761706

 
alfa aesar™
A1761706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1761714

 
alfa aesar™
A1761714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 270.55
Assay ≥98%
Molecular Formula C7H2ClF3N2O4
MDL Number MFCD00007076
CAS 392-95-0
UN Number UN2923
Beilstein 2061156
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
RTECS Number CZ0525750
Melting Point 61° to 64°C
Quantity 5g
Chemical Name or Material 2-Chloro-3,5-dinitrobenzotrifluoride

Applications
It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

Solubility
Insoluble in water. Soluble in Chloroform, Methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.