missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3,5-dinitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1761714

 View more versions of this product

Catalog No. AAA1761714


Description

Description

It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to increase mutagenic activity in the glutathione transferase mutant (C36).

Solubility
Insoluble in water. Soluble in Chloroform, Methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.
Specifications

Specifications

2-Chloro-3,5-dinitrobenzotrifluoride
61°C to 64°C
MFCD00007076
UN2923
2-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,5-dinitro-3-trifluoromethyl benzene, benzene, 2-chloro-1,5-dinitro-3-trifluoromethyl, 2-chloro-3,5-binitrotrifluorotoluene, pubchem4846, 2-chlor-1,5-dinitro-3-trifluormethyl benzol, ksc914e2d, labotest-bb lt00159813, labotest-bb lt03380099, 2-chloro-3,5-dinitro-benzotrifluoride
RLXKADBMLQPLDV-UHFFFAOYSA-N
2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
67851
≥98%
392-95-0
C7H2ClF3N2O4
25 g
2061156
Insoluble in water. Soluble in Chloroform,Methanol.
[O-][N+](=O)C1=CC(=C(Cl)C(=C1)C(F)(F)F)[N+]([O-])=O
270.55
270.55

Chemical Identifiers

392-95-0
270.55
RLXKADBMLQPLDV-UHFFFAOYSA-N
67851
[O-][N+](=O)C1=CC(=C(Cl)C(=C1)C(F)(F)F)[N+]([O-])=O
C7H2ClF3N2O4
MFCD00007076
2-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,5-dinitro-3-trifluoromethyl benzene, benzene, 2-chloro-1,5-dinitro-3-trifluoromethyl, 2-chloro-3,5-binitrotrifluorotoluene, pubchem4846, 2-chlor-1,5-dinitro-3-trifluormethyl benzol, ksc914e2d, labotest-bb lt00159813, labotest-bb lt03380099, 2-chloro-3,5-dinitro-benzotrifluoride
2-chloro-1,5-dinitro-3-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 206-883-0

RTECSNumber : CZ0525750

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only