Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,5-dinitrobenzoic acid, 97% (dry wt.), may cont. up to ca 5% water

$33.76 - $357.11

Chemical Identifiers

Chemical Name or Material 2-Chloro-3,5-dinitrobenzoic acid
CAS 2497-91-8
Molecular Formula C7H3ClN2O6
Formula Weight 246.56
MDL Number MFCD00007070
Beilstein 2146480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1567514

  SDS
alfa aesar™
A1567514
25g Each for $33.76

AAA1567522

  SDS
alfa aesar™
A1567522
100g Each for $84.82

AAA1567536

  SDS
alfa aesar™
A1567536
500g Each for $357.11
Description & Specifications

Specifications

Formula Weight 246.56
Molecular Formula C7H3ClN2O6
Health Hazard 1 Warning
Melting Point 196° to 201°C
Assay 97%
Beilstein 2146480
CAS 2497-91-8
RTECS Number DG4996500
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes (dry wt.), may cont. up to ≈5% water
MDL Number MFCD00007070
Chemical Name or Material 2-Chloro-3,5-dinitrobenzoic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.