missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,5-dinitrobenzoic acid, 97% (dry wt.), may cont. up to ca 5% water

Manufacturer:  Alfa Aesar™ A1567536

 View more versions of this product

Catalog No. AAA1567536



Specifications

Specifications

2-Chloro-3,5-dinitrobenzoic acid
500g
(dry wt.), may cont. up to ≈5% water
MFCD00007070
2-chloro-3,5-dinitrobenzoic acid, benzoic acid, 2-chloro-3,5-dinitro, 2-carboxy-4,6-dinitrochlorobenzene, 3,5-dinitro-2-chlorobenzoic acid, acmc-209uud, timtec-bb sbb003177, wln: wnr bg cvq enw, 4-09-00-01359 beilstein handbook reference, ksc495o8b, 2-chloro-3,5-dinitro-benzoicaci
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)[N+](=O)[O-]
17247
97%
196°C to 201°C
2497-91-8
C7H3ClN2O6
2146480
ADTKEYLCJYYHHH-UHFFFAOYSA-N
246.559
246.56

Chemical Identifiers

2497-91-8
246.559
ADTKEYLCJYYHHH-UHFFFAOYSA-N
17247
C7H3ClN2O6
MFCD00007070
2-chloro-3,5-dinitrobenzoic acid, benzoic acid, 2-chloro-3,5-dinitro, 2-carboxy-4,6-dinitrochlorobenzene, 3,5-dinitro-2-chlorobenzoic acid, acmc-209uud, timtec-bb sbb003177, wln: wnr bg cvq enw, 4-09-00-01359 beilstein handbook reference, ksc495o8b, 2-chloro-3,5-dinitro-benzoicaci
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG4996500

Documents