missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3,5-difluorophenol, 98%, ACROS Organics™

Chemical Identifiers

CAS 206986-81-4
Molecular Formula C6H3ClF2O
Molecular Weight (g/mol) 164.54
InChI Key ZQWULOHSTAQTKJ-UHFFFAOYSA-N
Synonym 2-chloro-3,5-difluoro-phenol, 2-chloro-3,5-difluorophenol, 4,6-difluoro-2-hydroxychlorobenzene, acmc-1cs4q, phenol, 2-chloro-3,5-difluoro, phenol,2-chloro-3,5-difluoro
PubChem CID 2733297
IUPAC Name 2-chloro-3,5-difluorophenol
SMILES C1=C(C=C(C(=C1F)Cl)O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440380010
SDS View Documents
Thermo Fisher Scientific
AC440380010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440380050
SDS View Documents
Thermo Fisher Scientific
AC440380050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

206986-81-4
164.54
2-chloro-3,5-difluoro-phenol, 2-chloro-3,5-difluorophenol, 4,6-difluoro-2-hydroxychlorobenzene, acmc-1cs4q, phenol, 2-chloro-3,5-difluoro, phenol,2-chloro-3,5-difluoro
2-chloro-3,5-difluorophenol
C6H3ClF2O
ZQWULOHSTAQTKJ-UHFFFAOYSA-N
2733297
C1=C(C=C(C(=C1F)Cl)O)F

Specifications

66°C
Glass Bottle
White
1g
97.5
97.5% min. (GC)
2-chloro-3,5-difluoro-phenol, 2-chloro-3,5-difluorophenol, 4,6-difluoro-2-hydroxychlorobenzene, acmc-1cs4q, phenol, 2-chloro-3,5-difluoro, phenol,2-chloro-3,5-difluoro
C1=C(C=C(C(=C1F)Cl)O)F
164.54
164.54
98%
Authentic
76.0°C (37.0 mmHg)
45.0°C to 46.0°C
206986-81-4
100.0
C6H3ClF2O
ZQWULOHSTAQTKJ-UHFFFAOYSA-N
2-chloro-3,5-difluorophenol
2733297
Crystalline Powder
2-chloro-3, 5-difluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS