Learn More
2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6411914
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Chloro-3,5-bis(trifluoromethyl)aniline | |
Colorless to Yellow | |
C8H4ClF6N | |
MFCD04038716 | |
UN2810 | |
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine | |
JKUFETFGEJPHEM-UHFFFAOYSA-N | |
2-chloro-3,5-bis(trifluoromethyl)aniline | |
2757429 | |
98% |
201593-90-0 | |
1.595 g/mL | |
1.459 | |
25 g | |
13730810 | |
Slightly soluble in water. | |
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F | |
263.567 | |
263.57 | |
Liquid |
Chemical Identifiers
201593-90-0 | |
263.567 | |
JKUFETFGEJPHEM-UHFFFAOYSA-N | |
2757429 | |
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F |
C8H4ClF6N | |
MFCD04038716 | |
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine | |
2-chloro-3,5-bis(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures