missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%

Manufacturer:  Alfa Aesar™ H6411922

 View more versions of this product

Catalog No. AAH6411922



Description

Description

Applications
2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Chloro-3,5-bis(trifluoromethyl)aniline
1.4590
100g
C8H4ClF6N
UN2810
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F
263.567
263.57
98%
1.595g/mL
Colorless to Yellow
201593-90-0
MFCD04038716
13730810
JKUFETFGEJPHEM-UHFFFAOYSA-N
2-chloro-3,5-bis(trifluoromethyl)aniline
2757429
Liquid

Chemical Identifiers

201593-90-0
263.567
JKUFETFGEJPHEM-UHFFFAOYSA-N
2757429
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F
C8H4ClF6N
MFCD04038716
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
2-chloro-3,5-bis(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents