Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-3,4-dimethoxybenzyl alcohol, 97%

$47.61 - $183.82

Chemical Identifiers

Chemical Name or Material 2-Chloro-3,4-dimethoxybenzyl alcohol
CAS 93983-13-2
Molecular Formula C9H11ClO3
Formula Weight 202.64
MDL Number MFCD00210141
Synonym 2-Chloroveratryl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068803

  SDS
alfa aesar™
B2068803
1g Each for $47.61

AAB2068806

  SDS
alfa aesar™
B2068806
5g Each for $183.82
Description & Specifications

Specifications

Formula Weight 202.64
Molecular Formula C9H11ClO3
MDL Number MFCD00210141
CAS 93983-13-2
Quantity 1g
Synonym 2-Chloroveratryl alcohol
Melting Point 70° to 72°C
Assay 97%
Chemical Name or Material 2-Chloro-3,4-dimethoxybenzyl alcohol

Applications
It is applied in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Incompatible with strong oxidizing agents.