missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-3,4-dimethoxybenzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 5417-17-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9 H9 Cl O3
Linear Formula (CH3O)2C6H2(Cl)CHO
Formula Weight 200.62
MDL Number MFCD00084942
Synonym 2-Chloroveratraldehyde
Chemical Name or Material 2-Chloro-3, 4-dimethoxybenzaldehyde, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC365640250 SDS Thermo Fisher Scientific
AC365640250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5417-17-4
100.0
C9 H9 Cl O3
200.62
2-Chloroveratraldehyde
≥97.5%
97.5
98%
(CH3O)2C6H2(Cl)CHO
MFCD00084942
2-Chloro-3, 4-dimethoxybenzaldehyde, 98%

Specifications

Crystalline Powder
25g
69° to 73°C
Authentic
MFCD00084942
98%
Glass bottle
(CH3O)2C6H2(Cl)CHO
≥97.5%
200.62
Beige-yellow to beige
100.0
5417-17-4
C9 H9 Cl O3
97.5
2-Chloroveratraldehyde
2-Chloro-3, 4-dimethoxybenzaldehyde, 98%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.