Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2'-nitroacetanilide, 98%

$38.12 - $465.59

Chemical Identifiers

Chemical Name or Material 2-Chloro-2'-nitroacetanilide
CAS 10147-70-3
Molecular Formula C8H7ClN2O3
Formula Weight 214.61
MDL Number MFCD00024201
Synonym N-Chloroacetyl-2-nitroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1422106

 
alfa aesar™
A1422106
5g Each for $38.12

AAA1422114

 
alfa aesar™
A1422114
25g Each for $133.35

AAA1422122

 
alfa aesar™
A1422122
100g Each for $465.59
Description & Specifications

Specifications

Formula Weight 214.61
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 87° to 89°C
CAS 10147-70-3
Quantity 5g
Molecular Formula C8H7ClN2O3
Synonym N-Chloroacetyl-2-nitroaniline
MDL Number MFCD00024201
Health Hazard 1 Warning
Chemical Name or Material 2-Chloro-2'-nitroacetanilide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.