Learn More
2-Chloro-2-methylnonane, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04378406
Description
It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloro-2-methylnonane | |
Colorless | |
MFCD00060781 | |
UN3082 | |
QDJXQCKHBZEIJM-UHFFFAOYSA-N | |
2-chloro-2-methylnonane | |
138189 | |
≥90% |
4325-50-2 | |
C10H21Cl | |
5 g | |
dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl | |
CCCCCCCC(C)(C)Cl | |
176.728 | |
176.73 | |
Liquid |
Chemical Identifiers
4325-50-2 | |
176.728 | |
QDJXQCKHBZEIJM-UHFFFAOYSA-N | |
138189 | |
CCCCCCCC(C)(C)Cl |
C10H21Cl | |
MFCD00060781 | |
dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl | |
2-chloro-2-methylnonane |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H351
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only