Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2'-fluoro-3,3'-bipyridine, 95%

$145.70 - $521.34

Chemical Identifiers

Chemical Name or Material 2-Chloro-2'-fluoro-3,3'-bipyridine
CAS 942206-09-9
Molecular Formula C10H6ClFN2
Formula Weight 208.62
MDL Number MFCD09037427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26884MC

  SDS
alfa aesar™
H26884MC
100mg Each for $145.70

AAH26884ME

  SDS
alfa aesar™
H26884ME
500mg Each for $521.34
Description & Specifications

Specifications

Formula Weight 208.62
Chemical Name or Material 2-Chloro-2'-fluoro-3,3'-bipyridine
Assay 95%
Molecular Formula C10H6ClFN2
MDL Number MFCD09037427
CAS 942206-09-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100mg
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.