Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2',6'-diethylacetanilide, 96%

$79.27 - $306.31

Chemical Identifiers

Chemical Name or Material 2-Chloro-2',6'-diethylacetanilide
CAS 6967-29-9
Molecular Formula C12H16ClNO
Formula Weight 225.72
MDL Number MFCD00124800
Beilstein 2111384
Synonym N-Chloroacetyl-2,6-diethylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1313606

 
alfa aesar™
L1313606
5g Each for $79.27

AAL1313614

 
alfa aesar™
L1313614
25g Each for $306.31
Description & Specifications

Specifications

Formula Weight 225.72
Molecular Formula C12H16ClNO
MDL Number MFCD00124800
CAS 6967-29-9
Beilstein 2111384
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Chloro-2',6'-diethylacetanilide
Assay 96%
Synonym N-Chloroacetyl-2,6-diethylaniline
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 128° to 132°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.