Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2',4'-difluoroacetophenone, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-2',4'-difluoroacetophenone
CAS 51336-94-8
Molecular Formula C8H5ClF2O
Formula Weight 190.58
MDL Number MFCD00013252
UN Number UN3261
Synonym 2,4-Difluorophenacyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2248706

 
alfa aesar™
B2248706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2248714

 
alfa aesar™
B2248714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.58
Chemical Name or Material 2-Chloro-2',4'-difluoroacetophenone
Assay 98%
Molecular Formula C8H5ClF2O
MDL Number MFCD00013252
CAS 51336-94-8
UN Number UN3261
Synonym 2,4-Difluorophenacyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Melting Point 44° to 48°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.