Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2,2-difluoroacetophenone, 96%

$45.81 - $107.59

Chemical Identifiers

Chemical Name or Material 2-Chloro-2,2-difluoroacetophenone
CAS 384-67-8
Molecular Formula C8H5ClF2O
Formula Weight 190.58
MDL Number MFCD00042123
Beilstein 2251448
Synonym alpha,alpha-Difluorophenacyl chloride; alpha-Chloro-alpha,alpha-difluoroacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1696303

  SDS
alfa aesar™
L1696303
1g Each for $45.81

AAL1696306

  SDS
alfa aesar™
L1696306
5g Each for $107.59
Description & Specifications

Specifications

Formula Weight 190.58
CAS 384-67-8
Chemical Name or Material 2-Chloro-2,2-difluoroacetophenone
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym alpha,alpha-Difluorophenacyl chloride; alpha-Chloro-alpha,alpha-difluoroacetophenone
Molecular Formula C8H5ClF2O
MDL Number MFCD00042123
Boiling Point 95° to 97°C (37mmHg)
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Beilstein 2251448
Flash Point 72°C (161°F)
Assay 96%

Applications
Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.