missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-2,2-difluoroacetamide, 98%

$63.23 - $223.61

Chemical Identifiers

CAS 354-28-9
Molecular Formula C2H2ClF2NO
Molecular Weight (g/mol) 129.491
MDL Number MFCD00039775
InChI Key PSAKKOKLSDIKEK-UHFFFAOYSA-N
Synonym chlorodifluoroacetamide, acetamide, 2-chloro-2,2-difluoro, difluorochloroacetamide, snylgqdjptadtaetp@, acmc-20algv, acetamide,2-difluoro, pubchem14583, 2-chloranyl-2,2-bis fluoranyl ethanamide
PubChem CID 67715
IUPAC Name 2-chloro-2,2-difluoroacetamide
SMILES C(=O)(C(F)(F)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0078506 View Documents Alfa Aesar™
L0078506
5g Each for $63.23
 
AAL0078514 View Documents Alfa Aesar™
L0078514
25g Each for $223.61
 
Specifications

Chemical Identifiers

354-28-9
129.491
PSAKKOKLSDIKEK-UHFFFAOYSA-N
67715
C(=O)(C(F)(F)Cl)N
C2H2ClF2NO
MFCD00039775
chlorodifluoroacetamide, acetamide, 2-chloro-2,2-difluoro, difluorochloroacetamide, snylgqdjptadtaetp@, acmc-20algv, acetamide,2-difluoro, pubchem14583, 2-chloranyl-2,2-bis fluoranyl ethanamide
2-chloro-2,2-difluoroacetamide

Specifications

79°C to 84°C
354-28-9
MFCD00039775
chlorodifluoroacetamide, acetamide, 2-chloro-2,2-difluoro, difluorochloroacetamide, snylgqdjptadtaetp@, acmc-20algv, acetamide,2-difluoro, pubchem14583, 2-chloranyl-2,2-bis fluoranyl ethanamide
C(=O)(C(F)(F)Cl)N
129.491
129.49
2-Chloro-2,2-difluoroacetamide
5g
C2H2ClF2NO
1753808
PSAKKOKLSDIKEK-UHFFFAOYSA-N
2-chloro-2,2-difluoroacetamide
67715
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA