missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1H-benzimidazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4857-06-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
InChI Key AYPSHJCKSDNETA-UHFFFAOYSA-N
Synonym 1h-benzimidazole, 2-chloro, 2-chloro benzimidazole, 2-chloro-1h-1,3-benzimidazole, 2-chloro-1h-1,3-benzodiazole, 2-chloro-1h-benzo d imidazole, 2-chloro-1h-benzoimidazole, 2-chlorobenzimidazole, 2-chlorobenzoimidazole, benzimidazole, 2-chloro, pubchem7598
PubChem CID 78572
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291960250
SDS View Documents Promotion Details
Thermo Fisher Scientific
291960250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4857-06-1
152.58
1h-benzimidazole, 2-chloro, 2-chloro benzimidazole, 2-chloro-1h-1,3-benzimidazole, 2-chloro-1h-1,3-benzodiazole, 2-chloro-1h-benzo d imidazole, 2-chloro-1h-benzoimidazole, 2-chlorobenzimidazole, 2-chlorobenzoimidazole, benzimidazole, 2-chloro, pubchem7598
C7H5ClN2
AYPSHJCKSDNETA-UHFFFAOYSA-N
78572

Specifications

4857-06-1
100.0
200°C
Authentic
Glass bottle
25g
152.58
152.58
97%
96.0
Beige to Brown
97%
C7H5ClN2
1h-benzimidazole, 2-chloro, 2-chloro benzimidazole, 2-chloro-1h-1,3-benzimidazole, 2-chloro-1h-1,3-benzodiazole, 2-chloro-1h-benzo d imidazole, 2-chloro-1h-benzoimidazole, 2-chlorobenzimidazole, 2-chlorobenzoimidazole, benzimidazole, 2-chloro, pubchem7598
AYPSHJCKSDNETA-UHFFFAOYSA-N
78572
Crystalline Powder
2-Chloro-1H-benzimidazole
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-453-3

SDS
Product Certifications

RUO – Research Use Only