Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1-tritylimidazole, 97%

$75.57 - $213.96

Chemical Identifiers

Chemical Name or Material 2-Chloro-1-tritylimidazole
CAS 67478-48-2
Molecular Formula C22H17ClN2
Formula Weight 344.84
MDL Number MFCD02179527
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5186103

 
alfa aesar™
H5186103
1g Each for $213.96

AAH51861MD

 
alfa aesar™
H51861MD
250mg Each for $75.57
Description & Specifications

Specifications

Formula Weight 344.84
Chemical Name or Material 2-Chloro-1-tritylimidazole
Assay 97%
Molecular Formula C22H17ClN2
MDL Number MFCD02179527
CAS 67478-48-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 200° to 202°C
Quantity 1g

Applications
2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.