Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2-Chloro-1-phenylethanol, 97%

Chemical Identifiers

Chemical Name or Material (+/-)-2-Chloro-1-phenylethanol
CAS 1674-30-2
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD01075658
UN Number UN2810
Beilstein 907322
Synonym alpha-(Chloromethyl)benzyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1453803

  SDS
alfa aesar™
L1453803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1453806

  SDS
alfa aesar™
L1453806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.61
Chemical Name or Material (+/-)-2-Chloro-1-phenylethanol
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
MDL Number MFCD01075658
Synonym alpha-(Chloromethyl)benzyl alcohol
CAS 1674-30-2
UN Number UN2810
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Quantity 1g
Health Hazard 1 Warning
Molecular Formula C8H9ClO
Beilstein 907322
Flash Point >110°C (230°F)
Boiling Point 110° to 112°C (5mmHg)

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.