Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1-methylimidazole-5-boronic acid pinacol ester, 95%

$370.66 - $370.66

Chemical Identifiers

Chemical Name or Material 2-Chloro-1-methylimidazole-5-boronic acid pinacol ester
CAS 1315281-29-8
Molecular Formula C10H16BClN2O2
Formula Weight 242.52
MDL Number MFCD11100959
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31692MD

  SDS
alfa aesar™
H31692MD
250mg Each for $370.66
Description & Specifications

Specifications

Formula Weight 242.52
Chemical Name or Material 2-Chloro-1-methylimidazole-5-boronic acid pinacol ester
Melting Point 131° to 135°C
Assay 95%
Quantity 250mg
Molecular Formula C10H16BClN2O2
MDL Number MFCD11100959
CAS 1315281-29-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Solubility
Sparingly soluble in water (0.23 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C