missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1-methylbenzimidazole, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6355903

 View more versions of this product

Catalog No. AAH6355903



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-1-methylbenzimidazole
White to Yellow
MFCD00769821
2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
CN1C(Cl)=NC2=CC=CC=C12
166.61
166.61
1849-02-1
C8H7ClN2
1 g
UXZYKSFMGDWHGJ-UHFFFAOYSA-N
2-chloro-1-methylbenzimidazole
663080
Solid

Chemical Identifiers

1849-02-1
166.61
UXZYKSFMGDWHGJ-UHFFFAOYSA-N
663080
CN1C(Cl)=NC2=CC=CC=C12
C8H7ClN2
MFCD00769821
2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
2-chloro-1-methylbenzimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only