missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1-methylbenzimidazole

Manufacturer:  Alfa Aesar™ H6355906

 View more versions of this product

Catalog No. AAH6355906



Specifications

Specifications

2-Chloro-1-methylbenzimidazole
5g
C8H7ClN2
2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
CN1C2=CC=CC=C2N=C1Cl
166.608
166.61
White to Yellow
1849-02-1
MFCD00769821
UXZYKSFMGDWHGJ-UHFFFAOYSA-N
2-chloro-1-methylbenzimidazole
663080
Solid

Chemical Identifiers

1849-02-1
166.608
UXZYKSFMGDWHGJ-UHFFFAOYSA-N
663080
CN1C2=CC=CC=C2N=C1Cl
C8H7ClN2
MFCD00769821
2-chloro-1-methyl-1h-benzo d imidazole, 2-chloro-1-methyl-1h-benzimidazole, 2-chloro-1-methyl-1h-benzoimidazole, 2-chloro-1-methyl-1,3-benzodiazole, 1-methyl-1h-benzo d imidazol-2-yl chloride, 2-chloro-1-methyl-1h-1,3-benzodiazole, 1-methyl-2-chlorobenzimidazole, 2-chloro-1-methyl-benzimidazole, 1h-benzimidazole, 2-chloro-1-methyl, acmc-209em7
2-chloro-1-methylbenzimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents