Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1-fluoro-4-nitrobenzene, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-1-fluoro-4-nitrobenzene
CAS 350-30-1
Molecular Formula C6H3ClFNO2
Formula Weight 175.55
MDL Number MFCD00007206
UN Number UN2811
Beilstein 1944997
Synonym 3-Chloro-4-fluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1618314

 
alfa aesar™
A1618314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1618318

 
alfa aesar™
A1618318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1618336

 
alfa aesar™
A1618336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.55
Chemical Name or Material 2-Chloro-1-fluoro-4-nitrobenzene
Odor Characteristic
Assay 98%
Molecular Formula C6H3ClFNO2
MDL Number MFCD00007206
CAS 350-30-1
UN Number UN2811
Beilstein 1944997
Synonym 3-Chloro-4-fluoronitrobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number CZ0720000
Flash Point 112°C (233°F)
Boiling Point 227° to 232°C
Melting Point 42° to 45°C
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.