Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1,4-difluorobenzene, 98%

Chemical Identifiers

Chemical Name or Material 2-Chloro-1,4-difluorobenzene
CAS 2367-91-1
Molecular Formula C6H3ClF2
Formula Weight 148.54
MDL Number MFCD00042571
UN Number UN1993
Beilstein 2081082
Synonym 1-Chloro-2,5-difluorobenzene; 2,5-Difluorochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1417806

 
alfa aesar™
A1417806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1417814

 
alfa aesar™
A1417814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1417822

 
alfa aesar™
A1417822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.54
Chemical Name or Material 2-Chloro-1,4-difluorobenzene
Odor Aromatic
Assay 98%
Molecular Formula C6H3ClF2
MDL Number MFCD00042571
CAS 2367-91-1
UN Number UN1993
Beilstein 2081082
Synonym 1-Chloro-2,5-difluorobenzene; 2,5-Difluorochlorobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 20°C (68°F)
Boiling Point 126° to 128°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P261-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.