Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1,4-di-n-propoxybenzene, 97%

$50.18 - $712.69

Chemical Identifiers

Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene
CAS 916791-98-5
Molecular Formula C12H17ClO2
Formula Weight 228.72
MDL Number MFCD00048688
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2477609

 
alfa aesar™
B2477609
10g Each for $50.18

AAB2477618

 
alfa aesar™
B2477618
50g Each for $177.01

AAB2477630

 
alfa aesar™
B2477630
250g Each for $712.69
Description & Specifications

Specifications

Formula Weight 228.72
Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene
Assay 97%
Molecular Formula C12H17ClO2
MDL Number MFCD00048688
CAS 916791-98-5
Quantity 10g