Please login to your online account to display your discounted pricing

2-Chloro-1,3-thiazole-5-carboxylic acid, 97%, Maybridge


Quantity 5g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC62901DE

 View more versions of this product

Catalog No. CC62901DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 163.58
Quantity 5g
Assay 97%
Molecular Formula C4H2ClNO2S
Assay Percent Range 97%
Packaging Amber glass bottle
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carboxylic acid
CAS 101012-12-8
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H2ClNO2S
Formula Weight 163.58
Accessories

Warning