missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1,3-thiazole-5-carbaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 95453-58-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H2ClNOS
Formula Weight 147.58
MDL Number MFCD01568717
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carbaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC62904CB SDS Fisher Scientific Company
CC62904CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62904DA SDS Fisher Scientific Company
CC62904DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95453-58-0
100.0
C4H2ClNOS
MFCD01568717
97%
96.5
97%
147.58
2-Chloro-1, 3-thiazole-5-carbaldehyde

Specifications

250mg
96.5
Amber Glass Bottle
C4H2ClNOS
MFCD01568717
97%
95453-58-0
100.0
97%
147.58
2-Chloro-1, 3-thiazole-5-carbaldehyde

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning