missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439460050

 View more versions of this product

Catalog No. AC439460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

2-chloro-1, 3-dimethylimidazolidinium tetrafluoroborate
White
5g
95.0
95% min. (HPLC)
CIB
220.41
96%
Authentic
58.0°C to 60.0°C
153433-26-2
100.0
C5H10ClN2·BF4
220.41
Powder or Crystalline Powder

Chemical Identifiers

153433-26-2
C5H10ClN2·BF4
220.41
CIB
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents