Please login to your online account to display your discounted pricing

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-chloro-1, 3-dimethylimidazolidinium tetrafluoroborate
CAS 153433-26-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C5H10BClF4N2
Formula Weight 220.41
Synonym CIB
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439460010

 
thermo fisher scientific
AC439460010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439460050

 
thermo fisher scientific
AC439460050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥95.0%
Formula Weight 220.41
Quantity 1g
Color White to off-white
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolidinium tetrafluoroborate
Assay Percent Range 96%
Molecular Formula C5H10BClF4N2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 153433-26-2
Synonym CIB
Infrared Spectrum Authentic

Coupling reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.