missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloro-1,3-difluoro-4-nitrobenzene, TCI America™

Manufacturer:  TCI America C296425G

 View more versions of this product

Catalog No. C296425G



Specifications

Specifications

2-Chloro-1,3-difluoro-4-nitrobenzene
47°C
3847-58-3
MFCD00042201
3-chloro-2,4-difluoronitrobenzene, 2,4-difluoro-3-chloronitrobenzene, benzene, 2-chloro-1,3-difluoro-4-nitro, 2,4-difluoro-3-chloronitrobnzene, 3-chloro-2,4-difluoro-1-nitrobenzene, 2-chloro-1,3-difluoro-4-nitro-benzene, pubchem4320, acmc-209j0b, ksc911k8t, buttpark 177\01-33
C1=CC(=C(C(=C1[N+](=O)[O-])F)Cl)F
193.534
193.53
≥98.0% (GC)
Yellow
25g
C6H2ClF2NO2
1759
CTVBVNKEMCGZDF-UHFFFAOYSA-N
2-chloro-1,3-difluoro-4-nitrobenzene
223080
Crystal-Powder at 20°C

Chemical Identifiers

3847-58-3
193.534
CTVBVNKEMCGZDF-UHFFFAOYSA-N
223080
C1=CC(=C(C(=C1[N+](=O)[O-])F)Cl)F
C6H2ClF2NO2
MFCD00042201
3-chloro-2,4-difluoronitrobenzene, 2,4-difluoro-3-chloronitrobenzene, benzene, 2-chloro-1,3-difluoro-4-nitro, 2,4-difluoro-3-chloronitrobnzene, 3-chloro-2,4-difluoro-1-nitrobenzene, 2-chloro-1,3-difluoro-4-nitro-benzene, pubchem4320, acmc-209j0b, ksc911k8t, buttpark 177\01-33
2-chloro-1,3-difluoro-4-nitrobenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents