Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1-(2,4-dichlorophenoxy)-4-nitrobenzene, 98%

$1,417.43 - $1,417.43

Chemical Identifiers

Chemical Name or Material 2-Chloro-1-(2,4-dichlorophenoxy)-4-nitrobenzene
CAS 22544-04-3
Molecular Formula C12H6Cl3NO3
Formula Weight 318.54g/mol
MDL Number MFCD00204155
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2516814

 
alfa aesar™
B25168-14
25g Each for $1,417.43
Description & Specifications

Specifications

Melting Point 70° to 72°C
Quantity 25g
Chemical Name or Material 2-Chloro-1-(2,4-dichlorophenoxy)-4-nitrobenzene
CAS 22544-04-3
Assay 98%
Molecular Formula C12H6Cl3NO3
Formula Weight 318.54g/mol
MDL Number MFCD00204155
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.