missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether, 97%

Manufacturer:  Alfa Aesar™ L1672514

 View more versions of this product

Catalog No. AAL1672514



Specifications

Specifications

2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether
25g
C3H2ClF5O
MFCD00069095
14,3581
JPGQOUSTVILISH-UHFFFAOYSA-N
2-chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane
3226
184.49
56°C to 57°C
13838-16-9
CHF2OCF2CHClF
1737129
2-chloro-1,1,2-trifluoroethyl difluoromethyl ether
C(C(OC(F)F)(F)F)(F)Cl
184.49
CHEBI:4792
97%

Chemical Identifiers

13838-16-9
184.49
JPGQOUSTVILISH-UHFFFAOYSA-N
3226
2-chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane
C3H2ClF5O
MFCD00069095
2-chloro-1,1,2-trifluoroethyl difluoromethyl ether
CHEBI:4792
C(C(OC(F)F)(F)F)(F)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : KN6800000

Recommended Storage : 2° to 8°C

Documents