Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Carboxyphenyl phosphate, 98%

$118.99 - $2,256.58

Chemical Identifiers

Chemical Name or Material 2-Carboxyphenyl phosphate
CAS 6064-83-1
Molecular Formula C7H7O6P
Formula Weight 218.1
MDL Number MFCD00042645
Beilstein 2619142
Merck Index 14,4255
Synonym Fosfosal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1441703

  SDS
alfa aesar™
A1441703
1g Each for $118.99

AAA1441706

  SDS
alfa aesar™
A1441706
5g Each for $535.68

AAA1441714

  SDS
alfa aesar™
A1441714
25g Each for $2,256.58
Description & Specifications

Specifications

Formula Weight 218.1
Chemical Name or Material 2-Carboxyphenyl phosphate
MDL Number MFCD00042645
CAS 6064-83-1
Beilstein 2619142
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point 162° to 166°C
Quantity 1g
Assay 98%
Molecular Formula C7H7O6P
Merck Index 14,4255
Synonym Fosfosal
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VO2770000

Applications
2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.