Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Carboxyphenyl)iminodiacetic acid, 96%

$36.61 - $412.85

Chemical Identifiers

Chemical Name or Material (2-Carboxyphenyl)iminodiacetic acid
CAS 1147-65-5
Molecular Formula C11H11NO6
Formula Weight 253.21
MDL Number MFCD00021751
Beilstein 2815076
Synonym N,N-Bis(carboxymethyl)anthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1291403

  SDS
alfa aesar™
L1291403
1g Each for $36.61

AAL1291406

  SDS
alfa aesar™
L1291406
5g Each for $106.68

AAL1291414

  SDS
alfa aesar™
L1291414
25g Each for $412.85
Description & Specifications

Specifications

Formula Weight 253.21
Chemical Name or Material (2-Carboxyphenyl)iminodiacetic acid
Molecular Formula C11H11NO6
MDL Number MFCD00021751
CAS 1147-65-5
Beilstein 2815076
Synonym N,N-Bis(carboxymethyl)anthranilic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 215° to 219°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.