missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Carboxyphenyl)iminodiacetic acid, 96%

Manufacturer:  Alfa Aesar™ L1291414

 View more versions of this product

Catalog No. AAL1291414



Specifications

Specifications

(2-Carboxyphenyl)iminodiacetic acid
25g
C11H11NO6
2815076
IKEUZCGOPYSHNN-UHFFFAOYSA-N
2-[bis(carboxymethyl)amino]benzoic acid
459325
96%
215°C to 219°C
1147-65-5
MFCD00021751
2-bis carboxymethyl amino benzoic acid, 2-carboxyphenyl iminodiacetic acid, 2,2'-2-carboxyphenyl azanediyl diacetic acid, 2-carboxyphenyliminodiacetic acid, acmc-20appp, n-2-carboxyphenyl iminodiacetic acid, n,n-bis carboxymethyl anthranilic acid, benzoic acid, 2-bis carboxymethyl amino
C1=CC=C(C(=C1)C(=O)O)N(CC(=O)O)CC(=O)O
253.21
253.21

Chemical Identifiers

1147-65-5
253.21
IKEUZCGOPYSHNN-UHFFFAOYSA-N
459325
C1=CC=C(C(=C1)C(=O)O)N(CC(=O)O)CC(=O)O
C11H11NO6
MFCD00021751
2-bis carboxymethyl amino benzoic acid, 2-carboxyphenyl iminodiacetic acid, 2,2'-2-carboxyphenyl azanediyl diacetic acid, 2-carboxyphenyliminodiacetic acid, acmc-20appp, n-2-carboxyphenyl iminodiacetic acid, n,n-bis carboxymethyl anthranilic acid, benzoic acid, 2-bis carboxymethyl amino
2-[bis(carboxymethyl)amino]benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents