missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Carboxymethylthio)pyrimidine, 98%

$173.93 - $173.93

Chemical Identifiers

CAS 88768-45-0
Molecular Formula C6H6N2O2S
Formula Weight 170.19
MDL Number MFCD00010287
Beilstein 136468
Synonym (2-Pyrimidinylthio)acetic acid; 2-(Carboxymethylmercapto)pyrimidine
Chemical Name or Material 2- (Carboxymethylthio)pyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1065914 SDS Alfa Aesar™
A1065914
25g Each for $173.93
AAA1065906 SDS Alfa Aesar™
A1065906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

88768-45-0
170.19
136468
2- (Carboxymethylthio)pyrimidine
C6H6N2O2S
MFCD00010287
(2-Pyrimidinylthio)acetic acid; 2-(Carboxymethylmercapto)pyrimidine

Specifications

≈200°C (dec.)
88768-45-0
170.19
136468
2- (Carboxymethylthio)pyrimidine
25g
C6H6N2O2S
MFCD00010287
(2-Pyrimidinylthio)acetic acid; 2-(Carboxymethylmercapto)pyrimidine
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning