Learn More
(2-Carboxyethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
$103.64 - $968.07
Chemical Identifiers
CAS | 36626-29-6 |
---|---|
Molecular Formula | C21H20ClO2P |
Molecular Weight (g/mol) | 370.81 |
MDL Number | MFCD00185663 |
InChI Key | GALLWJZTZYJVSL-UHFFFAOYSA-N |
Synonym | 2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride |
PubChem CID | 10270534 |
SMILES | [Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2560906
|
Thermo Scientific Chemicals
B2560906 |
5 g |
Each for $103.64
|
|
|||||
AAB2560914
|
Thermo Scientific Chemicals
B2560914 |
25 g |
Each for $335.82
|
|
|||||
AAB2560922
|
Thermo Scientific Chemicals
B2560922 |
100 g |
Each for $968.07
|
|
|||||
Description
2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Carboxyethyl)-triphenylphosphonium chloride is a phosphonium ylide reagent for Wittig olefinations with introduction of a carboxylic acid.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Chemical Identifiers
36626-29-6 | |
370.81 | |
GALLWJZTZYJVSL-UHFFFAOYSA-N | |
10270534 |
C21H20ClO2P | |
MFCD00185663 | |
2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride | |
[Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
36626-29-6 | |
C21H20ClO2P | |
5 g | |
2-carboxyethyl triphenylphosphonium chloride, 2-carboxyethyl tripheylphosphonium chloride, 2-carboxyethyl-triphenylphosphonium chloride, phosphonium, 2-carboxyethyl triphenyl-, chloride, 2-carboxyethyl triphenylphosphanium chloride, acmc-1agne, 2-carboxyethyl triphenyl phosphonium chloride, beta-carboxyethyltriphenylphosphonium chloride, 2-carboxyethyl triphenyl-phosphonium chloride | |
GALLWJZTZYJVSL-UHFFFAOYSA-N | |
2-carboxyethyl(triphenyl)phosphanium;chloride | |
10270534 | |
98% |
197°C to 201°C | |
MFCD00185663 | |
Hygroscopic | |
Soluble in dimethyl sulfoxide and methanol. | |
[Cl-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
370.81 | |
370.81 | |
(2-Carboxyethyl)triphenylphosphonium chloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only