Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Carboxy-3-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Carboxy-3-fluorobenzeneboronic acid pinacol ester
Formula Weight 266.07
Synonym 2-Carboxy-3-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH62754MD

 
alfa aesar™
H62754MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6275403

 
alfa aesar™
H6275403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.07
Quantity 250mg
Physical Form Solid
Assay 96%
Chemical Name or Material 2-Carboxy-3-fluorobenzeneboronic acid pinacol ester
Synonym 2-Carboxy-3-fluorophenylboronic acid pinacol ester