Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Carboxy-3,4-dimethoxybenzaldehyde, 99%

$77.91 - $277.24

Chemical Identifiers

Chemical Name or Material 2-Carboxy-3,4-dimethoxybenzaldehyde
CAS 519-05-1
Molecular Formula C10H10O5
Formula Weight 210.19
MDL Number MFCD00039572
Beilstein 2696087
Merck Index 14,6851
Synonym 5,6-Dimethoxyphthalaldehydic acid; 6-Formyl-2,3-dimethoxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1626306

  SDS
alfa aesar™
L1626306
5g Each for $277.24

AAL1626303

  SDS
alfa aesar™
L1626303
1g Each for $77.91
Description & Specifications

Specifications

Formula Weight 210.19
CAS 519-05-1
MDL Number MFCD00039572
Chemical Name or Material 2-Carboxy-3,4-dimethoxybenzaldehyde
Beilstein 2696087
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H10O5
Merck Index 14,6851
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Synonym 5,6-Dimethoxyphthalaldehydic acid; 6-Formyl-2,3-dimethoxybenzoic acid
Melting Point 145° to 148°C
Assay 99%

Applications
Anthraquinones, hydrazones has been prepared using 2-Carboxy-3,4-dimethoxybenzaldehyde.

Solubility
Soluble in water. 2.5 g/L (20°C).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Store at +15°C to +25°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.