Please login to your online account to display your discounted pricing

2-Carbomethoxy-3-thiophenesulfonyl chloride, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398610050

 View more versions of this product

Catalog No. AC398610050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥94%
Formula Weight 240.69
Quantity 5g
Color White to brown
Melting Point 60° to 65°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Carbomethoxy-3-thiophenesulfonyl chloride
Assay Percent Range 95%
MDL Number MFCD00068160
CAS 59337-92-7
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H5ClO4S2
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Methyle3-chlorosulfonylthiophene-2-carboxylate
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Carbomethoxy-3-thiophenesulfonyl chloride
CAS 59337-92-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5ClO4S2
Formula Weight 240.69
MDL Number MFCD00068160
Synonym Methyle3-chlorosulfonylthiophene-2-carboxylate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents