Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butyrylthiophene, 97%

$33.86 - $227.81

Chemical Identifiers

Chemical Name or Material 2-Butyrylthiophene
CAS 5333-83-5
Molecular Formula C8H10OS
Formula Weight 154.23
MDL Number MFCD00022503
Beilstein 110167
Synonym n-Propyl 2-thienyl ketone; 1-(2-Thienyl)-1-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1504806

 
alfa aesar™
A1504806
5g Each for $33.86

AAA1504809

 
alfa aesar™
A1504809
10g Each for $56.96

AAA1504818

 
alfa aesar™
A1504818
50g Each for $227.81
Description & Specifications

Specifications

Formula Weight 154.23
Chemical Name or Material 2-Butyrylthiophene
Assay 97%
Molecular Formula C8H10OS
MDL Number MFCD00022503
CAS 5333-83-5
Beilstein 110167
Synonym n-Propyl 2-thienyl ketone; 1-(2-Thienyl)-1-butanone
Boiling Point 242° to 244°C
Quantity 5g
Flash Point 102°C (216°F)
Odor Characteristic