missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butyrylthiophene, 97%

Manufacturer:  Alfa Aesar™ A1504818

 View more versions of this product

Catalog No. AAA1504818



Specifications

Specifications

2-Butyrylthiophene
Characteristic
50g
C8H10OS
110167
YXHIINNJOGKPLF-UHFFFAOYSA-N
1-thiophen-2-ylbutan-1-one
79248
97%
102°C (216°F)
242°C to 244°C
5333-83-5
MFCD00022503
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz
CCCC(=O)C1=CC=CS1
154.227
154.23

Chemical Identifiers

5333-83-5
154.227
YXHIINNJOGKPLF-UHFFFAOYSA-N
79248
CCCC(=O)C1=CC=CS1
C8H10OS
MFCD00022503
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz
1-thiophen-2-ylbutan-1-one
Documents