missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 2-Butyrylthiophene, 97%
Manufacturer: Alfa Aesar™ A1504809
Specifications
2-Butyrylthiophene | |
Characteristic | |
10g | |
C8H10OS | |
110167 | |
YXHIINNJOGKPLF-UHFFFAOYSA-N | |
1-thiophen-2-ylbutan-1-one | |
79248 | |
97% |
102°C (216°F) | |
242°C to 244°C | |
5333-83-5 | |
MFCD00022503 | |
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz | |
CCCC(=O)C1=CC=CS1 | |
154.227 | |
154.23 |
Chemical Identifiers
5333-83-5 | |
154.227 | |
YXHIINNJOGKPLF-UHFFFAOYSA-N | |
79248 | |
CCCC(=O)C1=CC=CS1 |
C8H10OS | |
MFCD00022503 | |
2-butyrylthiophene, 1-2-thienyl-1-butanone, 1-thiophen-2-yl butan-1-one, 2-n-butyrylthiophene, 2-butanoylthiophene, 1-thien-2-ylbutan-1-one, 1-2-thienyl butan-1-one, ketone, propyl 2-thienyl, 1-butanone, 1-2-thienyl, unii-biq5m88olz | |
1-thiophen-2-ylbutan-1-one |