Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butyrylfuran, 97%

$24.55 - $56.50

Chemical Identifiers

Chemical Name or Material 2-Butyrylfuran
CAS 4208-57-5
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00046111
Synonym 1-(2-Furyl)-1-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2628703

  SDS
alfa aesar™
H2628703
1g Each for $24.55

AAH2628706

  SDS
alfa aesar™
H2628706
5g Each for $56.50
Description & Specifications

Specifications

Formula Weight 138.17
Chemical Name or Material 2-Butyrylfuran
Assay 97%
Molecular Formula C8H10O2
MDL Number MFCD00046111
CAS 4208-57-5
Synonym 1-(2-Furyl)-1-butanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.